Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem196 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044269.1 | NP_001037734.1 | 3 | 747 | ||
XM_006240701.2 | XP_006240763.1 | 3 | 1991 | |||
XM_006240702.2 | XP_006240764.1 | 3 | 1991 | |||
XM_008765005.1 | XP_008763227.1 | 3 | 1973 | |||
GenBank mRNA | BC089997.1 | - | 3 | 747 | 7320460 | |
FQ211598.1 | - | 3 | 150 |