Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100995.1 | NP_001094465.1 | 3, 4 | 680 | ||
GenBank mRNA | BC129127.1 | - | 3, 4 | 680 | 7190631 | |
FQ224194.1 | - | 3, 4 | 718 |