Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Odf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191992.1 | NP_001178921.1 | 4 | 472 | ||
XM_006242237.1 | XP_006242299.1 | 4 | 581 | |||
XM_006242238.2 | XP_006242300.1 | 5 | 1115 | |||
XM_008765776.1 | XP_008763998.1 | 4 | 855 |