Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1563200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109288.1 | NP_001102758.1 | 3, 4 | 332 | ||
XM_008765777.1 | XP_008763999.1 | 3, 4 | 294 | |||
GenBank mRNA | BC167768.1 | - | 3, 4 | 325 | 7456523 |