Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134634.2 | NP_001128106.2 | 8 | 918 | ||
XM_006242578.2 | XP_006242640.1 | 7 | 910 | |||
XM_008765969.1 | XP_008764191.1 | 5 | 656 | |||
XM_008765970.1 | XP_008764192.1 | 6 | 717 | |||
XM_008765971.1 | XP_008764193.1 | 5 | 660 | |||
XM_008765972.1 | XP_008764194.1 | 6 | 721 | |||
XM_008765973.1 | XP_008764195.1 | 4 | 598 | |||
XM_008765974.1 | XP_008764196.1 | 4 | 602 | |||
XM_008765975.1 | XP_008764197.1 | 5 | 632 | |||
XM_008765976.1 | XP_008764198.1 | 5 | 562 | |||
XM_008765977.1 | XP_008764199.1 | 5 | 538 | |||
GenBank mRNA | BU671127.1 | - | 4 | 288 | ||
FQ211838.1 | - | 6 | 705 |