Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC500990 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037364.1 | NP_001032441.1 | 8 | 905 | ||
XM_008766163.1 | XP_008764385.1 | 9 | 1121 | |||
XM_008766164.1 | XP_008764386.1 | 9 | 1121 | |||
XM_008766165.1 | XP_008764387.1 | 9 | 1121 | |||
XM_008766166.1 | XP_008764388.1 | 9 | 1121 | |||
XM_008766167.1 | XP_008764389.1 | 9 | 1118 | |||
XM_008766168.1 | XP_008764390.1 | 9 | 1132 | |||
XM_008766169.1 | XP_008764391.1 | 8 | 1038 | |||
XM_008766170.1 | XP_008764392.1 | 8 | 993 | |||
GenBank mRNA | BC105843.1 | - | 8 | 905 | 7455643 |