Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Layn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191997.1 | NP_001178926.1 | 7 | 686 | ||
XM_006243049.2 | XP_006243111.1 | 6 | 1651 | |||
XM_006243052.2 | XP_006243114.1 | 5 | 535 | |||
XM_008766328.1 | XP_008764550.1 | 5 | 1615 |