Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Treml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192001.1 | NP_001178930.1 | 3, 4 | 615 | ||
GenBank mRNA | FQ227710.1 | - | 3, 4 | 625 |