Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdfi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109301.1 | NP_001102771.1 | 5 | 1028 | ||
XM_006244475.2 | XP_006244537.1 | 5 | 789 | |||
XM_006244476.2 | XP_006244538.1 | 5 | 982 | |||
XM_008766877.1 | XP_008765099.1 | 5 | 955 | |||
GenBank mRNA | BC158727.1 | - | 4 | 950 | 9091612 |