Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC501110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024361.1 | NP_001019532.1 | 6 | 614 | ||
XM_006244656.2 | XP_006244718.1 | 7 | 717 | |||
XM_006244657.2 | XP_006244719.1 | 7 | 684 | |||
XM_006244658.2 | XP_006244720.1 | 7 | 1066 | |||
GenBank mRNA | BC079271.1 | - | 6 | 614 | 7133989 |