Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Satb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109306.1 | NP_001102776.1 | 9 | 1691 | ||
XM_006244931.2 | XP_006244993.1 | 9 | 2363 | |||
XM_008767128.1 | XP_008765350.1 | 8 | 2135 | |||
XM_008767129.1 | XP_008765351.1 | 9 | 1637 |