Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110493.1 | NP_001103963.1 | 4 | 755 | ||
GenBank mRNA | AY325140.1 | - | 4 | 755 |