Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244357.2 | XP_006244419.1 | 12 | 1128 | ||
XM_006244358.2 | XP_006244420.1 | 11 | 1100 | |||
XM_006244359.2 | XP_006244421.1 | 9 | 942 | |||
XM_006244360.2 | XP_006244422.1 | 11 | 923 | |||
XM_006244361.2 | XP_006244423.1 | 12 | 1131 | |||
XM_006244362.2 | XP_006244424.1 | 8 | 737 | |||
XM_576696.6 | XP_576696.4 | 10 | 895 | |||
GenBank mRNA | FQ212208.1 | - | 11 | 961 |