Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126088.1 | NP_001119560.1 | 2, 3 | 228 | ||
XM_006227894.1 | XP_006227956.1 | 2, 3 | 189 | |||
GenBank mRNA | FQ212207.1 | - | 150 |