Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aplp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100802.1 | NP_001094272.1 | 17 | 1932 | ||
XM_008759179.1 | XP_008757401.1 | 16 | 1813 | |||
GenBank mRNA | BC161904.1 | - | 17 | 1932 | 8367333 | |
DQ462463.1 | - | 2 | 154 | |||
FQ212221.1 | - | 17 | 1922 | |||
FQ213451.1 | - | 17 | 1974 |