Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dok5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109344.2 | NP_001102814.1 | 5 | 538 | ||
XM_006235673.2 | XP_006235735.1 | 5 | 717 | |||
GenBank mRNA | BC167027.1 | - | 5 | 538 | 7315978 |