Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1561560 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235972.2 | XP_006236034.1 | 2 | 473 | ||
XM_578210.6 | XP_578210.4 | 2 | 328 |