Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC503435 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109366.1 | NP_001102836.1 | 1 | 609 | ||
XM_006256820.2 | XP_006256882.1 | 2 | 1300 | |||
XM_008773133.1 | XP_008771355.1 | 2 | 846 |