Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012649.1 | NP_001012667.1 | 6, 7 | 819 | ||
XM_008763328.1 | XP_008761550.1 | 5, 6 | 828 | |||
GenBank mRNA | AY286494.1 | - | 6, 7 | 819 | ||
FQ233804.1 | - | 6, 7 | 837 |