Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017359.2 | NP_059055.2 | 3, 4 | 373 | ||
GenBank mRNA | AY310140.1 | - | 1, 2 | 137 | ||
AY724493.1 | - | 3, 4 | 552 | |||
BC085744.1 | - | 3, 4 | 373 | 7191921 |