Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001053826.4 | XP_001053826.1 | 1 | 351 | ||
XM_006242286.2 | XP_006242348.1 | 1 | 351 |