Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255782.2 | XP_006255844.1 | 2 | 297 | ||
GenBank mRNA | BC088147.1 | - | 2 | 239 | 7323596 | |
U12268.1 | - | 2 | 202 |