Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc14a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110270.1 | NP_001103740.1 | 5, 6 | 1265 | ||
NM_019347.2 | NP_062220.2 | 16, 17 | 2309 | |||
GenBank mRNA | U09957.1 | - | 5, 6 | 1264 | ||
U77971.1 | - | 16, 17 | 2309 |