Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgals1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019904.1 | NP_063969.1 | 2 | 60 | ||
GenBank mRNA | BC058476.1 | - | 2 | 90 | 6920799 | |
FQ217640.1 | - | 2 | 87 | |||
FQ221017.1 | - | 2 | 86 | |||
FQ221731.1 | - | 2 | 86 | |||
FQ221808.1 | - | 2 | 87 | |||
FQ221945.1 | - | 2 | 87 | |||
FQ222079.1 | - | 2 | 87 | |||
FQ222712.1 | - | 2 | 87 | |||
FQ223159.1 | - | 2 | 86 | |||
FQ223277.1 | - | 2 | 87 | |||
FQ223897.1 | - | 2 | 87 | |||
FQ224769.1 | - | 2 | 87 | |||
FQ228416.1 | - | 1 | 14 | |||
FQ228452.1 | - | 2 | 87 | |||
FQ228459.1 | - | 2 | 86 | |||
FQ228622.1 | - | 2 | 85 | |||
FQ230068.1 | - | 2 | 87 | |||
M19036.1 | - | 2 | 60 |