Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Park2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020093.1 | NP_064478.1 | 10 | 1253 | ||
GenBank mRNA | AB039878.1 | - | 10 | 1253 | ||
AF168004.1 | - | 10 | 1273 | |||
AF210434.1 | - | 10 | 1245 | |||
AF257234.1 | - | 9 | 1183 | |||
AF343574.1 | - | 10 | 942 | |||
AF381277.1 | - | 10 | 1255 | |||
AF381278.1 | - | 9 | 1183 | |||
AF381280.1 | - | 10 | 1339 | |||
AF381281.1 | - | 8 | 1099 | |||
AF381285.1 | - | 3 | 319 | |||
GU322017.1 | - | 11 | 1352 | |||
GU322019.1 | - | 11 | 1283 | |||
GU322363.1 | - | 9 | 1112 | |||
GU345835.1 | - | 11 | 1430 | |||
GU345836.1 | - | 11 | 1231 | |||
KC774169.1 | - | 9 | 1039 | |||
KC774170.1 | - | 9 | 1039 | |||
KC774172.1 | - | 10 | 1193 | |||
KC774173.1 | - | 7 | 827 | |||
KC774174.1 | - | 5 | 476 | |||
KC774175.1 | - | 7 | 893 | |||
KC774176.1 | - | 7 | 840 | |||
KC774177.1 | - | 5 | 621 |