Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2afz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022674.1 | NP_073165.1 | 4 | 310 | ||
GenBank mRNA | BC060564.1 | - | 4 | 375 | 6922321 | |
BC086348.1 | - | 4 | 355 | 7302603 | ||
FQ213837.1 | - | 4 | 369 | |||
FQ220829.1 | - | 4 | 371 | |||
FQ221791.1 | - | 4 | 369 | |||
FQ222470.1 | - | 4 | 369 | |||
FQ224994.1 | - | 4 | 372 | |||
FQ227672.1 | - | 4 | 371 | |||
FQ229641.1 | - | 4 | 371 | |||
FQ229746.1 | - | 4 | 371 | |||
FQ232362.1 | - | 4 | 371 | |||
FQ233167.1 | - | 4 | 371 | |||
FQ234498.1 | - | 4 | 371 | |||
M37584.1 | - | 4 | 310 | |||
X52316.1 | - | 4 | 281 |