Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabarap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172036.3 | NP_742033.1 | 3 | 238 | ||
GenBank mRNA | AF161588.1 | - | 3 | 252 | ||
BC058441.1 | - | 3 | 242 | 6918795 | ||
FQ213199.1 | - | 3 | 309 | |||
FQ224284.1 | - | 3 | 204 | |||
FQ229256.1 | - | 3 | 240 |