Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ensa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033974.2 | NP_001029146.1 | 1, 2 | 191 | ||
NM_021842.3 | NP_068614.1 | 1, 2 | 191 | |||
GenBank mRNA | AJ005984.1 | - | 1, 2 | 51 | ||
BC059135.1 | - | 1, 2 | 195 | 6919652 | ||
CB812628.1 | - | 1, 2 | 170 | |||
FQ212889.1 | - | 1, 2 | 156 | |||
FQ212980.1 | - | 1, 2 | 173 | |||
FQ213096.1 | - | 1, 2 | 158 | |||
FQ213912.1 | - | 1, 2 | 175 | |||
FQ214207.1 | - | 1, 2 | 169 | |||
FQ216114.1 | - | 1, 2 | 175 | |||
FQ216323.1 | - | 1, 2 | 150 |