Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ensa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033974.2 | NP_001029146.1 | 3 | 387 | ||
NM_021842.3 | NP_068614.1 | 3 | 387 | |||
GenBank mRNA | AJ005984.1 | - | 3 | 247 | ||
BC059135.1 | - | 3 | 391 | 6919652 | ||
CB812628.1 | - | 3 | 366 | |||
FQ212889.1 | - | 3 | 352 | |||
FQ212892.1 | - | 3 | 371 | |||
FQ212980.1 | - | 3 | 369 | |||
FQ213096.1 | - | 3 | 354 | |||
FQ213912.1 | - | 3 | 371 | |||
FQ214207.1 | - | 3 | 365 | |||
FQ216114.1 | - | 3 | 371 | |||
FQ216323.1 | - | 3 | 346 |