Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031660.1 | NP_113848.1 | 2 | 63 | ||
XM_006243410.2 | XP_006243472.1 | 3 | 174 | |||
GenBank mRNA | AJ005982.1 | - | 2 | 63 | ||
BC058461.1 | - | 3 | 109 | 6919944 | ||
FQ213926.1 | - | 1 | 144 | |||
FQ221182.1 | - | 3 | 113 | |||
FQ222843.1 | - | 3 | 112 |
Gene Symbol: LOC100360828 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729972.4 | XP_002730018.2 | 1 | 100 |