Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037292.1 | NP_001032369.1 | 152 | |||
GenBank mRNA | U13176.1 | - | 152 |