Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uba52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031687.3 | NP_113875.1 | 4 | 309 | ||
XM_006252934.2 | XP_006252996.1 | 4 | 396 | |||
XM_006252935.1 | XP_006252997.1 | 4 | 514 | |||
XM_008771145.1 | XP_008769367.1 | 4 | 392 | |||
XM_008771146.1 | XP_008769368.1 | 4 | 336 | |||
GenBank mRNA | BC061544.1 | - | 4 | 305 | 6922034 | |
BP480615.1 | - | 1 | 171 | |||
BP492585.1 | - | 3 | 195 | |||
FQ210541.1 | - | 4 | 298 | |||
FQ210904.1 | - | 4 | 299 | |||
FQ212001.1 | - | 4 | 303 | |||
FQ212205.1 | - | 4 | 301 | |||
FQ212291.1 | - | 4 | 299 | |||
FQ212810.1 | - | 4 | 302 | |||
FQ217153.1 | - | 4 | 310 | |||
FQ217440.1 | - | 4 | 294 | |||
FQ219376.1 | - | 4 | 300 | |||
FQ219803.1 | - | 4 | 301 | |||
FQ221045.1 | - | 4 | 298 | |||
FQ221118.1 | - | 4 | 299 | |||
FQ221238.1 | - | 4 | 519 | |||
FQ221621.1 | - | 4 | 295 | |||
FQ221631.1 | - | 4 | 301 | |||
FQ221755.1 | - | 4 | 299 | |||
FQ221772.1 | - | 4 | 299 | |||
FQ221777.1 | - | 4 | 299 | |||
FQ222045.1 | - | 4 | 299 | |||
FQ222098.1 | - | 4 | 295 | |||
FQ222371.1 | - | 4 | 298 | |||
FQ222440.1 | - | 4 | 310 | |||
FQ222483.1 | - | 4 | 298 | |||
FQ222507.1 | - | 4 | 293 | |||
FQ222550.1 | - | 4 | 295 | |||
FQ222700.1 | - | 4 | 298 | |||
FQ223258.1 | - | 4 | 301 | |||
FQ224647.1 | - | 3 | 289 | |||
FQ224702.1 | - | 4 | 299 | |||
FQ228362.1 | - | 4 | 295 | |||
FQ228566.1 | - | 4 | 299 | |||
U25064.1 | - | 1 | 32 | |||
X82636.1 | - | 3 | 271 |