Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ascl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022384.1 | NP_071779.1 | 1 | 1324 | ||
GenBank mRNA | X53725.1 | - | 1 | 1324 |