Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPLP0 SIRNA Assays ›
Gene Symbol:
Rplp0
Gene Name:
ribosomal protein, large, P0
Gene Aliases:
Arbp
Chromosome Location:
Chr.12: 46791528 - 46794797 on Build Rnor_6.0
UniGene ID:
Rn.1079
Species:
Rat
siRNA ID
RSS368058
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rplp0
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022402.2
NP_071797.1
5
628
GenBank mRNA
BC062028.1
-
4
597
5623881
BP465362.1
-
3
462
DN931898.1
-
4
646
FQ209537.1
-
5
628
FQ209603.1
-
5
628
FQ209620.1
-
4
613
FQ209757.1
-
5
629
FQ210253.1
-
5
628
FQ210412.1
-
5
628
FQ211152.1
-
5
628
FQ212554.1
-
5
628
FQ212743.1
-
5
628
FQ212875.1
-
5
628
FQ212927.1
-
5
629
FQ213085.1
-
5
628
FQ213427.1
-
5
628
FQ213510.1
-
5
628
FQ213712.1
-
5
628
FQ213847.1
-
5
629
FQ214087.1
-
5
628
FQ214515.1
-
5
628
FQ214553.1
-
5
629
FQ214850.1
-
5
628
FQ214997.1
-
5
628
FQ215007.1
-
5
628
FQ215548.1
-
5
630
FQ215662.1
-
5
628
FQ215682.1
-
5
628
FQ215870.1
-
5
628
FQ216120.1
-
5
629
FQ216296.1
-
5
628
FQ216389.1
-
5
629
FQ216406.1
-
5
631
FQ216495.1
-
5
628
FQ216618.1
-
5
628
FQ216655.1
-
5
629
FQ216733.1
-
5
630
FQ216774.1
-
5
628
FQ216813.1
-
5
627
FQ216851.1
-
5
628
FQ216913.1
-
5
629
FQ217406.1
-
5
628
FQ218561.1
-
5
628
FQ218832.1
-
5
628
FQ218943.1
-
5
628
FQ219025.1
-
5
631
FQ219184.1
-
5
632
FQ219389.1
-
5
628
FQ219457.1
-
5
628
FQ219678.1
-
5
628
FQ219815.1
-
5
628
FQ219817.1
-
5
628
FQ219837.1
-
5
628
FQ219855.1
-
5
628
FQ219923.1
-
5
628
FQ220008.1
-
5
628
FQ220051.1
-
5
628
FQ220077.1
-
5
628
FQ220115.1
-
5
628
FQ220181.1
-
5
628
FQ220201.1
-
5
626
FQ220213.1
-
5
628
FQ220247.1
-
5
629
FQ220282.1
-
5
630
FQ220284.1
-
5
628
FQ220302.1
-
5
628
FQ220338.1
-
5
628
FQ220345.1
-
5
629
FQ220370.1
-
5
628
FQ220406.1
-
5
628
FQ220413.1
-
5
628
FQ220420.1
-
5
629
FQ220444.1
-
5
628
FQ220445.1
-
5
624
FQ220449.1
-
5
628
FQ220514.1
-
5
628
FQ220527.1
-
5
628
FQ220533.1
-
5
660
FQ220542.1
-
5
628
FQ220545.1
-
5
628
FQ220634.1
-
5
628
FQ220654.1
-
5
628
FQ220663.1
-
5
628
FQ220684.1
-
5
629
FQ220769.1
-
5
628
FQ220787.1
-
5
628
FQ220801.1
-
5
628
FQ220833.1
-
5
628
FQ220838.1
-
5
628
FQ220888.1
-
5
628
FQ220932.1
-
5
629
FQ221146.1
-
5
628
FQ221380.1
-
5
628
FQ221556.1
-
5
626
FQ222171.1
-
5
628
FQ222181.1
-
5
628
FQ222327.1
-
5
628
FQ222772.1
-
5
628
FQ222906.1
-
5
628
FQ223121.1
-
5
628
FQ223487.1
-
5
628
FQ224547.1
-
5
628
FQ224834.1
-
5
628
FQ224846.1
-
5
628
FQ225049.1
-
5
628
FQ225170.1
-
5
628
FQ225608.1
-
5
628
FQ225654.1
-
5
630
FQ225860.1
-
5
630
FQ225886.1
-
5
628
FQ226306.1
-
5
630
FQ227381.1
-
5
628
FQ227522.1
-
5
623
FQ227719.1
-
5
628
FQ227837.1
-
5
628
FQ227937.1
-
5
629
FQ227941.1
-
5
628
FQ227942.1
-
5
748
FQ228548.1
-
5
628
FQ228995.1
-
5
628
FQ228996.1
-
5
628
FQ229139.1
-
5
630
FQ229159.1
-
5
628
FQ229164.1
-
5
629
FQ229208.1
-
5
628
FQ229241.1
-
5
628
FQ229244.1
-
5
628
FQ229248.1
-
3
508
FQ229272.1
-
5
628
FQ229273.1
-
5
629
FQ229303.1
-
5
628
FQ229337.1
-
5
628
FQ229352.1
-
5
628
FQ229365.1
-
5
628
FQ229397.1
-
5
628
FQ229408.1
-
5
629
FQ229433.1
-
5
628
FQ229448.1
-
5
628
FQ229521.1
-
5
628
FQ229547.1
-
5
628
FQ229554.1
-
5
628
FQ229566.1
-
5
624
FQ229610.1
-
5
628
FQ229626.1
-
5
625
FQ229726.1
-
5
628
FQ229740.1
-
5
628
FQ229742.1
-
5
628
FQ229776.1
-
5
628
FQ229808.1
-
5
628
FQ229815.1
-
5
628
FQ229822.1
-
5
628
FQ229830.1
-
5
630
FQ229883.1
-
5
630
FQ229929.1
-
5
628
FQ229932.1
-
3
526
FQ229952.1
-
5
628
FQ229974.1
-
5
628
FQ230005.1
-
5
628
FQ230055.1
-
5
627
FQ230127.1
-
5
628
FQ230267.1
-
5
631
FQ230943.1
-
5
628
FQ231445.1
-
5
630
FQ231525.1
-
5
628
FQ231591.1
-
5
630
FQ231710.1
-
5
629
FQ231733.1
-
5
629
FQ231897.1
-
5
630
FQ231967.1
-
5
628
FQ232029.1
-
5
629
FQ232154.1
-
5
628
FQ232165.1
-
5
628
FQ232310.1
-
5
627
FQ232319.1
-
5
629
FQ232697.1
-
5
630
FQ232767.1
-
5
628
FQ232951.1
-
5
628
FQ233152.1
-
5
627
FQ233377.1
-
5
628
FQ233473.1
-
5
628
FQ233732.1
-
5
628
FQ233737.1
-
5
628
FQ233808.1
-
5
628
FQ234195.1
-
5
628
FQ234341.1
-
5
628
FQ234613.1
-
5
628
FQ234733.1
-
5
628
FQ234735.1
-
5
518
FQ234863.1
-
5
628
FQ235063.1
-
5
628
FQ235079.1
-
5
628
FQ235259.1
-
5
628
X15096.1
-
5
628
Z29530.1
-
4
584
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational elongation
ribosome biogenesis
cellular response to interleukin-4
structural constituent of ribosome
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7745c474b-dptlh:80/100.66.78.226:80.
git-commit: c3c9db9cc82507548abdd0ae6a4ade3f81f66292
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.20.0-2024.11.45-1.0