Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022508.1 | NP_071953.1 | 13, 14 | 1607 | ||
XM_006240249.2 | XP_006240311.1 | 13, 14 | 1746 | |||
GenBank mRNA | BC089800.1 | - | 13, 14 | 1321 | 7323085 | |
J05519.1 | - | 13, 14 | 1607 |