Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022542.1 | NP_071987.1 | 1 | 300 | ||
GenBank mRNA | M74295.1 | - | 1 | 300 |