Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022544.2 | NP_071989.1 | 2 | 237 | ||
GenBank mRNA | AF068861.1 | - | 2 | 123 | ||
AY621374.1 | - | 2 | 237 |