Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022548.1 | NP_071993.1 | 2 | 445 | ||
GenBank mRNA | Y13148.1 | - | 2 | 445 |