Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031695.2 | NP_113883.2 | 4, 5 | 824 | ||
XM_006255625.2 | XP_006255687.1 | 6, 7 | 1999 | |||
XM_006255626.2 | XP_006255688.1 | 7, 8 | 2191 | |||
XM_006255627.2 | XP_006255689.1 | 5, 6 | 1881 | |||
GenBank mRNA | CO404237.1 | - | 2, 3 | 214 | ||
X76988.1 | - | 836 |