Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cited1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172055.2 | NP_742052.2 | 3 | 449 | ||
XM_006257097.2 | XP_006257159.1 | 3 | 574 | |||
XM_006257098.2 | XP_006257160.1 | 4 | 1138 | |||
XM_006257099.2 | XP_006257161.1 | 3 | 1020 | |||
XM_006257100.2 | XP_006257162.1 | 3 | 464 | |||
GenBank mRNA | AF104399.1 | - | 2 | 307 | ||
BC097276.1 | - | 3 | 449 | 7446633 |