Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022598.1 | NP_072120.1 | 4, 5 | 589 | ||
XM_006236854.2 | XP_006236916.1 | 4, 5 | 584 | |||
GenBank mRNA | AB158421.1 | - | 3, 4 | 424 | ||
AB158422.1 | - | 3, 4 | 424 | |||
AF242550.1 | - | 4, 5 | 475 | |||
BC062225.1 | - | 4, 5 | 573 | 6889682 | ||
D45254.1 | - | 4, 5 | 589 |