Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hes3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022687.1 | NP_073178.1 | 2, 3 | 351 | ||
XM_008764381.1 | XP_008762603.1 | 3, 4 | 448 | |||
XM_008764382.1 | XP_008762604.1 | 3, 4 | 384 | |||
XM_008764383.1 | XP_008762605.1 | 3, 4 | 326 | |||
GenBank mRNA | D13418.1 | - | 2, 3 | 351 |