Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270838.1 | NP_001257767.1 | 9 | 1149 | ||
NM_001270839.1 | NP_001257768.1 | 9 | 1127 | |||
NM_001270840.1 | NP_001257769.1 | 9 | 1146 | |||
NM_022941.4 | NP_075230.1 | 9 | 1131 | |||
XM_006250261.2 | XP_006250323.1 | 9 | 1237 | |||
XM_008769759.1 | XP_008767981.1 | 8 | 2094 | |||
GenBank mRNA | AB104736.1 | - | 9 | 1184 | ||
AB105072.1 | - | 8 | 1275 | |||
AB204897.1 | - | 8 | 997 | |||
AB204898.1 | - | 8 | 1000 | |||
AB204899.1 | - | 8 | 978 | |||
AB204900.1 | - | 7 | 1177 | |||
AF133094.1 | - | 8 | 982 | |||
AF133095.1 | - | 8 | 982 | |||
BC089763.1 | - | 9 | 1146 | 7324318 |
Gene Symbol: Tomm40l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006250247.2 | XP_006250309.1 | 10 | 2420 |