Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ruvbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147177.1 | NP_671706.1 | 183 | |||
XM_006236865.2 | XP_006236927.1 | 257 | ||||
GenBank mRNA | AB001581.1 | - | 170 | |||
AB002406.1 | - | 183 | ||||
BC072511.1 | - | 168 | 7104777 | |||
BC086531.1 | - | 201 | 7304521 |