Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001053214.4 | XP_001053214.2 | 14 | 3105 | ||
XM_006228482.1 | XP_006228544.1 | 14 | 3277 | |||
XM_006228483.2 | XP_006228545.1 | 15 | 3814 | |||
XM_006228484.2 | XP_006228546.1 | 15 | 3168 | |||
XM_006228485.2 | XP_006228547.1 | 15 | 3497 | |||
XM_006228486.2 | XP_006228548.1 | 14 | 3669 | |||
XM_006228487.1 | XP_006228549.1 | 14 | 3028 | |||
XM_006228488.2 | XP_006228550.1 | 15 | 3380 | |||
XM_006228489.1 | XP_006228551.1 | 15 | 3167 | |||
XM_006228491.2 | XP_006228553.1 | 15 | 3291 | |||
XM_006228493.1 | XP_006228555.1 | 13 | 3028 | |||
XM_008759031.1 | XP_008757253.1 | 14 | 3222 |