Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam72a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081451.1 | NP_001074920.1 | 2 | 195 | ||
GenBank mRNA | AY513720.1 | - | 2 | 195 |