Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001131032.1 | NP_001124504.1 | 2 | 933 | ||
GenBank mRNA | EU176857.1 | - | 2 | 933 |