Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101005.1 | NP_001094475.1 | 5 | 601 | ||
XM_006235253.2 | XP_006235315.1 | 4 | 594 | |||
GenBank mRNA | BC083575.1 | - | 5 | 601 | 7112877 | |
FQ209572.1 | - | 4 | 554 | |||
FQ225908.1 | - | 4 | 491 |