Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127636.1 | NP_001121108.1 | 5 | 495 | ||
XM_008770571.1 | XP_008768793.1 | 4 | 442 | |||
GenBank mRNA | BC166528.1 | - | 5 | 495 | 7189304 |