Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102552318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760184.1 | XP_008758406.1 | 5 | 780 | ||
XM_008760185.1 | XP_008758407.1 | 5 | 779 |